missing translation for 'onlineSavingsMsg'
Learn More

Salmonella Rabbit anti-Bacteria, FITC, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA173020

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11578391

  • £340.00 / 1mL

Estimated Shipment date
from Supplier 01-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications

Specifications

Salmonella
Polyclonal
FITC
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Immunofluorescence
4-5 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide
Salmonellae
Mixture of S. enteriditis, S. typhimurium & S. heidelburg (O & H antigens).
Purified
RUO
Antibody
Polyclonal
Videos
Documents