Learn More
S100P Mouse anti-Human, Unlabeled, Clone: 16, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610306
Additional Details : Weight : 0.01000kg
Description
The S100 multigene family consists of a number of small dimeric molecules around 100 residues in length. The original members of this family, S100α and S100β, were initially identified in bovine brain. S100P is a novel member of this family which was isolated from human placenta. The S100 family consists of at least 10 members which show a cell-type-specific expression pattern. Most S100 proteins are believed to be involved in cytoplasmic Ca2+ control. S100P consists of 95 amino acids and shares about 50% sequence identity with S100α and S100α. Like other S100 proteins, S100P functions as a homodimer and binds two Ca2+ ions per monomer. Each S100P molecule contains both a low affinity and a high affinity Ca2+ binding site.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
S100P | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
16 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.