missing translation for 'onlineSavingsMsg'
Learn More

RT1B Mouse anti-Rat, FITC, Clone: OX-6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 554928

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10176083

  • £499.00 / Each

Estimated Shipment: 08-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The OX-6 antibody reacts with non-polymorphic determinants of the Rat MHC class II antigen, I-A equivalent. RT1B is found on peripheral B lymphocytes, thymic cortical epithelial and medullary reticular cells, small intestinal villus epithelium, epidermal Langerhans cells, dendritic cells, some tissue macrophage populations, peritoneal mast cells, and a subset of thymocytes, but not on peripheral T cells, erythrocytes, or microglia. The OX-6 mAb cross-reacts with mouse I-A[k] and I-A[s] alloantigens and with a major subset of splenocytes from NOD (I-A[g7]) mice.

Flow Cytometry

Specifications

Specifications

RT1B
OX-6
FITC
Affinity Purified
Ia-like Glycoproteins from Wistar Thymocytes
0.5mg
Immunology
Primary
Murine, Rat
Flow Cytometry
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.