missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RT1.B Monoclonal Antibody (OX-6), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517428
147.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
RT1.B Monoclonal antibody specifically detects RT1.B in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
RT1.B | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
0 | |
RT1-B | |
Rat thymocyte membrane glycoprotein | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
OX-6 | |
Biotin | |
RT1-B | |
rat class I B; RT1 class II, locus B; RT1-B | |
Mouse | |
Protein G | |
RUO | |
24738 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |