missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RPS6KB2 Matched Antibody Pair, Abnova™
Brand: Abnova H00006199-AP51.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-RPS6KB2 (100μg); Detection antibody: rabbit purified polyclonal anti-RPS6KB2 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RPS6KB2.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
RPS6KB2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RPS6KB2. | |
KLS/P70-beta/P70-beta-1/P70-beta-2/S6K-beta2/S6K2/SRK/STK14B/p70(S6K)-beta/p70S6Kb | |
Mouse, Rabbit | |
RUO | |
6199 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
RPS6KB2 | |
RPS6KB2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-RPS6KB2 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-RPS6KB2 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |