Learn More
ROBO1 Mouse anti-Rat, Clone: 243706, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB17491
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448885
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
ROBO1 Monoclonal antibody MAB17491 Specifically detects ROBO1 in Rat samples. It is validated for ELISA Capture
Specifications
Specifications
ROBO1
243706
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ROBO1
Mouse myeloma cell line NS0-derived recombinant rat ROBO1 Lys19-Ile560 Accession # O55005
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6091
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O55005
DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat ROBO1 in direct ELISAs and Western blots.
Rat