missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASET2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532031
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNASET2 Polyclonal antibody specifically detects RNASET2 in Human samples. It is validated for Western Blot.Specifications
RNASET2 | |
Polyclonal | |
Unconjugated | |
RNASET2 | |
0610007O07Rik; 4833423A10Rik; 4930532K22Rik; bA514O12.3; C79750; Ribonuclease 6; Ribonuclease 6-A; Ribonuclease 6-B; Ribonuclease T2; ribonuclease T2A; Ribonuclease T2-A; Ribonuclease T2-B; RNase T2; RNASE6PL; RNASET2; Rnaset2a; Rnaset2b; rnt2; RP11-514O12.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O00584 | |
RNASET2 | |
Recombinant fragment corresponding to a region within amino acids 26 and 256 of Human Ribonuclease T2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |