missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530397
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (91%), Rat (90%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RGS14 Polyclonal antibody specifically detects RGS14 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RGS14 | |
Polyclonal | |
Unconjugated | |
Rgs14 | |
rap1/rap2 interacting protein; RAP1/RAP2-interacting protein; Regulator of G-protein signaling 14; regulator of G-protein signalling 14; RGS14; RGSE; RPIP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10636, 51791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43566, P97492 | |
Rgs14 | |
Recombinant fragment corresponding to a region within amino acids 1 and 193 of Human RGS14 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |