Learn More
RGM-B Mouse anti-Human, Clone: 398528, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3630
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450176
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
RGM-B Monoclonal antibody MAB3630 Specifically detects RGM-B in Human samples. It is validated for Western Blotting
Specifications
Specifications
RGM-B
398528
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
RGMB
Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-Ser411 Accession # Q6NW40
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
285704
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q6NW40
DKFZp434P228, Dragon, DRG11-responsive axonal guidance and outgrowth of neurite, RGM domain family, member B, RGMB
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human RGM-B in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse RGM-B, recombinant human (rh) RGM-A, or rhRGM-C is observed.