Learn More
RELM gamma Mouse anti-Rat, Clone: 395310, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2789
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449695
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
RELM gamma Monoclonal antibody MAB2789 Specifically detects RELM gamma in Rat samples. It is validated for Western Blotting
Specifications
Specifications
RELM gamma
395310
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse myeloma cell line NS0-derived recombinant rat RELM gamma Asp24-Ser111 Accession # NP_853656
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects rat RELM gamma in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Resistin, recombinant mouse (rm) Resistin, rmRELM alpha , rmRELM beta , or rhRELM beta is observed.
Rat
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_853656
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.