Learn More
Pro Relaxin-3 Mouse anti-Human, Clone: 332112, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3107
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449895
  • £288.60 / 100µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
Pro Relaxin-3 Monoclonal antibody MAB3107 Specifically detects Pro Relaxin-3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Relaxin-3
332112
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
RLN3
E. coli-derived recombinant human Relaxin-3 Arg26-Cys142 Accession # Q8WXF3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
117579
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q8WXF3
H3, INSL7, M3, relaxin 3, Relaxin3, RLN3, RXN3, ZINS4
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Pro Relaxin-3 in direct ELISAs and Western blots. No cross-reactivity with mature human Relaxin-3, human Pro Relaxin-1 or human Pro Relaxin-2 is observed.
Human
Videos
missing translation for 'documents'
For Research Use Only