Learn More
Relaxin-1 Rat anti-Human, Clone: 361505, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB32571
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439975
  • £249.49 / 100µg
Estimated Shipment
27-06-2019
Description
Description
Relaxin-1 Monoclonal antibody MAB32571 Specifically detects Relaxin-1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Relaxin-1
361505
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
RLN1
E. coli-derived recombinant human Relaxin-1 Lys26-Cys185 Accession # P04808
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6013
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P04808
bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, Relaxin1, RLN1, RLXH1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects both the mature form and the proform of human Relaxin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Relaxin-2 or rhRelaxin-3 is observed.