missing translation for 'onlineSavingsMsg'
Learn More

Reg3D Rat anti-Mouse, Clone: 518818, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB5678

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461026

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Reg3D Monoclonal antibody MAB5678 Specifically detects Reg3D in Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

Reg3D
518818
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Reg3d
E. coli-derived recombinant mouse Reg3D, Glu27-Gly175
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
This antibody detects mouse Reg3D in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Reg1A, 1B, recombinant mouse (rm) Reg1, 2, 3A, 3B, 3G, 4, or recombinant rat Reg3 is observed.
Mouse
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
Purified
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only