Learn More
SMAGP Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3967
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488464
  • £382.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
SMAGP Polyclonal antibody AF3967 Specifically detects SMAGP in Rat samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
SMAGP
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant rat SMAGP, Met1-Thr34, Phe56-Ile97, Accession No. Q7TPF1
100μg
Primary
300236
Unconjugated
Q7TPF1
Goat
IgG
RUO
Polyclonal
Rat
Videos