missing translation for 'onlineSavingsMsg'
Learn More

Pentraxin 2/SAP Goat anti-Rat, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1895

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437214

  • £298.33 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pentraxin 2/SAP Polyclonal antibody AF1895 Specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Western Blotting
Specifications

Specifications

Pentraxin 2/SAP
LYOPH
Western Blot 0.1 ug/mL
P23680
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat Pentraxin 2/SAP in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human Pentraxin 2 and recombinant mouse Pentraxin 2 is observed.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
APCS
Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 Accession # P23680
Immunogen affinity purified
RUO
Primary
325
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only