Learn More
Neurexophilin-1 Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4208
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458614
  • £314.17 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Neurexophilin-1 Polyclonal antibody AF4208 Specifically detects Neurexophilin-1 in Rat samples. It is validated for Western Blotting
Specifications
Specifications
Neurexophilin-1
LYOPH
Western Blot 0.1 ug/mL
Q63366
neurexophilin 1, neurexophilin-1, Neurexophilin1, NPH1, NXPH1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat Neurexophilin-1/NXPH-1 in direct ELISAs and Western blots.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NXPH1
Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-1/NXPH-1 Ala22-Gly271 Accession # Q63366
Immunogen affinity purified
RUO
Primary
30010
Reconstitute at 0.2 mg/mL in sterile PBS.