Learn More
Goat anti-Rat IgG2c Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184711
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody may cross-react with IgG2c from other species. Goat anti Rat IgG2c antibody recognizes rat IgG2c as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgG2c antibody may cross react with IgG2c from other species.
Rat IgG2c Polyclonal antibody specifically detects Rat IgG2c in Rat samples. It is validated for ELISA, Western Blot.Specifications
Rat IgG2c | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |