Learn More
Goat anti-Rat IgG2b Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184710
Code : LD
Additional Details : Weight : 0.01000kg
Description
Goat anti Rat IgG2b antibody recognizes rat IgG2b as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgG2b antibody may cross react with IgG2b from other species.
Rat IgG2b Polyclonal antibody specifically detects Rat IgG2b in Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Rat IgG2b | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |