Learn More
Goat anti-Rat IgG2a Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184755
215.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
The antibody has been shown to react specifically with rat IgG2a by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG2a from other species. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Goat anti Rat IgG2a antibody recognizes rat IgG2a as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgG2a antibody may cross react with IgG2a from other species.
Rat IgG2a Polyclonal antibody specifically detects Rat IgG2a in Rat samples. It is validated for ELISA, Flow Cytometry, Western Blot.Specifications
Rat IgG2a | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Flow Cytometry, Western Blot | |
1 mg/mL | |
PBS with <0.1% sodium azide; pH 7.2 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |