Learn More
Goat anti-Rat IgG2a Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184761
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody has not been preabsorbed, and therefore may cross-react with IgG2a from other species. For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Goat anti Rat IgG2a antibody recognizes rat IgG2a as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgG2a antibody may cross react with IgG2a from other species.
Rat IgG2a Polyclonal antibody specifically detects Rat IgG2a in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Rat IgG2a | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA and <0.1% sodium azide | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |