Learn More
Rat IgG2a Isotype Control, FITC, Invitrogen™
Rat Isotype Control
Brand: Invitrogen PA533215
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PA5-33215 has been shown to be negative on all mouse cells and cell lines tested. This antibody recognizes a human cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that PA5-33215 is also suitable for use as a negative control with canine tissues.
Rat IgG2a Control antibody specifically detects Rat IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2a | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Rat | |
Protein G | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
FITC | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Human lymphocytes. | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG2a |