Learn More
Goat anti-Rat IgG1 Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184708
277.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody may cross-react with IgG1 from other species. Goat anti Rat IgG1 has been shown to react with rat IgG1 by immunoelectrophoresis and ELISA. Goat anti Rat IgG1 may cross react with IgG1 from other species.
Rat IgG1 Polyclonal antibody specifically detects Rat IgG1 in Rat samples. It is validated for ELISA, Western Blot.Specifications
Rat IgG1 | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |