missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Rat IgG1, κ, Brilliant Violet 786, Clone: R3-34, BD
Brand: BD Biosciences 563847
Additional Details : Weight : 0.02000kg
Description
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Immunogen: Mouse immunoglobulin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
R3-34 | |
Brilliant Violet 786 | |
Rat | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse immunoglobulin | |
50 μg | |
Isotype Control (Primary) | |
IgG1 κ |