Learn More
F(ab')2-Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™
F(ab')2-Donkey Polyclonal Secondary Antibody
Brand: Invitrogen A24543
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Rehydrate with 0.55 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rat serum proteins, IgG from bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit or sheep.
Rat IgG (H+L) Highly Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Highly Cross-Adsorbed in Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Rat IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
HRP | |
Donkey | |
Affinity Chromatography | |
RUO | |
Rat | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |