Learn More
Mouse anti-Rat IgE Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA516813
653.36 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.
Rat IgE Monoclonal antibody specifically detects Rat IgE in Rat samples. It is validated for ELISA, Western Blot.Specifications
Rat IgE | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal | |
Rat IgE myeloma protein (IR162, IR410) | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA, Western Blot | |
MARE-1 | |
HRP | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |