missing translation for 'onlineSavingsMsg'
Learn More

CCL26-like/Eotaxin-3-like Goat anti-Rat, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3878

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438424

  • £362.60 / 100µg

Estimated Shipment date
from Supplier 10-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CCL26-like/Eotaxin-3-like Polyclonal antibody AF3878 Specifically detects CCL26-like/Eotaxin-3-like in Rat samples. It is validated for Western Blotting
Specifications

Specifications

CCL26-like/Eotaxin-3-like
LYOPH
Western Blot 0.1 ug/mL
NP_001102958
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat CCL26-like/Eotaxin-3-like in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse CCL26-like is observed.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
E. coli-derived recombinant rat CCL26-like/Eotaxin-3-like Thr24-Trp94 Accession # NP_001102958
Immunogen affinity purified
RUO
Primary
2911
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only