Learn More
B7-2/CD86 Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1340
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13486884
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
B7-2/CD86 Polyclonal antibody AF1340 Specifically detects B7-2/CD86 in Rat samples. It is validated for ELISA Capture, Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications
Specifications
B7-2/CD86
Unconjugated
NP_064466
Goat
IgG
RUO
Polyclonal
Rat
Western Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant rat B7-2/CD86, Met1-Ile250, Accession No. NP_064466
100μg
Primary
56822
Videos