Learn More
B7-1/CD80 Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1214
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13486814
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
B7-1/CD80 Polyclonal antibody AF1214 Specifically detects B7-1/CD80 in Rat samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
B7-1/CD80
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80, Ile39-Gln248, Accession # AAB60503
100μg
Primary
25408
Unconjugated
AAB60503
Goat
IgG
RUO
Polyclonal
Rat
Videos