Learn More
ALK-7 Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF577
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429424
  • £342.51 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
ALK-7 Polyclonal antibody AF577 Specifically detects ALK-7 in Rat samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
ALK-7/Activin Receptor Type 1C
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
NP_620790
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat ALK-7 in direct ELISAs and Western blots.
Rat
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ACVR1C
Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 (predicted) Accession # NP_620790
Immunogen affinity purified
RUO
Primary
130399
Reconstitute at 0.2 mg/mL in sterile PBS.