missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527503
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji, K562. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RASSF7 Polyclonal antibody specifically detects RASSF7 in Human samples. It is validated for Western Blot.Specifications
RASSF7 | |
Polyclonal | |
Unconjugated | |
RASSF7 | |
2400009B11Rik; AW210608; C11orf13; HRAS1; HRAS1-related cluster protein 1; HRAS1-related cluster-1; HRC1; LOW QUALITY PROTEIN: ras association domain-containing protein 7; MGC126069; MGC126070; Ras association (RalGDS/AF-6) domain family (N-terminal) member 7; Ras association (RalGDS/AF-6) domain family 7; Ras association domain family member 7; ras association domain-containing protein 7; Rassf7; RGD1306244 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8045 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q02833 | |
RASSF7 | |
Recombinant fragment corresponding to a region within amino acids 146 and 373 of Human RASSF7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |