Learn More
RAP2C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521960
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (92%), Xenopus laevis (96%), Chicken (96%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAP2C Polyclonal antibody specifically detects RAP2C in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RAP2C | |
Polyclonal | |
Unconjugated | |
RAP2C | |
2010200P20Rik; AI194294; AL022976; RAP2C; RAP2C, member of RAS oncogene family; ras-related protein Rap-2c; small GTPase RAP2C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y3L5 | |
RAP2C | |
Full length human RAP2C Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |