Learn More
N2DL4 Monoclonal Antibody (205001), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523892
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant mouse Rae-1 alpha, beta, gamma, or gamma is observed. By flow cytometry, no cross-reactivity with mouse Rae-1 alpha or mouse Rae-1 gamma has been observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
N2DL4 Monoclonal antibody specifically detects N2DL4 in Mouse samples. It is validated for Flow Cytometry, Neutralization.Specifications
N2DL4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9CZQ6 | |
RAET1E | |
BaF3 mouse pro-B cell line transfected with mouse Rae-1epsilon | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Neutralization | |
205001 | |
Unconjugated | |
RAET1E | |
bA350J20.7; LETAL; Lymphocyte effector toxicity activation ligand; MGC125309; N2DL4; N2DL-4; NKG2D ligand 4; NKG2DL4; Rae-1 epsilon; RAE1 like transcript 4; RAE1epsilon; RAE-1-epsilon; RAE-1-like transcript 4; RAET1E; RAET1E2; Retinoic acid early transcript 1E; retinoic acid early-inducible protein 1-epsilon; RL-4; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4; ULBP4MGC125308; UNQ1867/PRO4303 | |
Rat | |
Protein A/G | |
RUO | |
379043 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |