Learn More
Rae-1 α/β/δ/ε Rat anti-Mouse, Clone: 199205, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB17581
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13468895
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Rae-1 α/β/δ/ε Monoclonal antibody MAB17581 Specifically detects Rae-1 α/β/δ/ε in Mouse samples. It is validated for Blocking (Receptor-ligand), Flow Cytometry
Specifications
Specifications
Rae-1
199205
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Raet1a
BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58)
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8480
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Purified
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Rae-1 alpha/beta/delta/epsilon . When tested in flow cytometry against a panel of transfectants expressing different Rae-1 proteins, this antibody strongly recognizes Rae-1 beta and Rae-1 delta . It shows weaker staining of Rae-1 alpha and Rae-1 epsilon transfectants. It does not recognize Rae-1 gamma .