missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Radixin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518542
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
Radixin Polyclonal antibody specifically detects Radixin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Radixin | |
Polyclonal | |
Unconjugated | |
Rdx | |
AA516625; DFNB24; ESP10; radixin; radixin isoform; Rdx | |
Goat | |
Antigen affinity chromatography | |
RUO | |
19684, 315655, 5962 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P26043, P35241 | |
Rdx | |
Synthetic peptide sequence (QARDETKKTQND) corresponding to the internal amino acids of RDX | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |