Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A16098
60.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Rabbit IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |