missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537209
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 42 kDa. Purity is >95% by SDS-PAGE.
RAB8B Polyclonal antibody specifically detects RAB8B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RAB8B | |
Polyclonal | |
Unconjugated | |
RAB8B | |
5930437D16; D330025I23Rik; FLJ38125; GTPase Rab8b; LOW QUALITY PROTEIN: ras-related protein Rab-8B; RAB44, member RAS oncogene family; RAB8B; RAB-8b protein; RAB8B, member RAS oncogene family; rab8bb; ras-related protein Rab-8B; ras-related protein Rab-8B; LOW QUALITY PROTEIN: ras-related protein Rab-8B; Ras-related protein Rab-8B-like protein; Unknown (protein for MGC:127591); zgc:165529 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
235442, 266688, 51762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P61028, P70550, Q92930 | |
RAB8B | |
A synthetic peptide corresponding to residues in human RAB8B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |