missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 3 Rat anti-Human, Mouse, Clone: 400403, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB41201

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440416

  • £286.65 / 100µg

Estimated Shipment date
from Supplier 24-01-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

R-Spondin 3 Monoclonal antibody MAB41201 Specifically detects R-Spondin 3 in Human, Mouse samples. It is validated for Western Blotting
Specifications

Specifications

R-Spondin 3
400403
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
RSPO3
E. coli-derived recombinant mouse R-Spondin 3 Ser21-Gly209 (Gln108Lys) Accession # BAC36296
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84870
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
BAC36296
Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse R-Spondin 3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) R-Spondin 1, recombinant human R-Spondin 2, or rmR-Spondin 4 is observed.
Human, Mouse
Videos
Documents

For Research Use Only