Learn More
PYY Polyclonal Antibody, Biotin, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA19590
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-9590 detects PYY in human, mouse and rat samples. PA1-9590 has been successfully used in ELISA procedures. PA1-9590 immunogen is a synthetic peptide corresponding to amino acids 3-18 (I K P E A P G E D A S P E E L N) of human PYY.
PYY Polyclonal antibody specifically detects PYY in Human, Mouse, Rat samples. It is validated for ELISA.Specifications
PYY | |
Polyclonal | |
Biotin | |
Pyy | |
GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; PYY; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy | |
Chicken | |
Affinity chromatography | |
RUO | |
217212, 287730, 5697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P10082, P10631, Q9EPS2 | |
Pyy | |
Synthetic peptide corresponding to amino acids 3-18 of human PYY. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |