Learn More
Nectin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530837
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (92%), Rat (94%), Dog (94%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Nectin 4 Polyclonal antibody specifically detects Nectin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Nectin 4 | |
Polyclonal | |
Unconjugated | |
NECTIN4 | |
1200017F15Rik; EDSS1; ig superfamily receptor LNIR; LNIR; nectin 4; nectin cell adhesion molecule 4; NECTIN4; Nectin-4; poliovirus receptor-related 4; poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4; PRR4; Pvrl4; RGD1559826 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q96NY8 | |
NECTIN4 | |
Recombinant fragment corresponding to a region within amino acids 29 and 300 of Human PVRL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |