Learn More
PSGL-1/CD162 Mouse anti-Human, Clone: CHO131, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB996
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13442086
  • £364.33 / 500µg
Estimated Shipment
08-07-2019
Description
Description
PSGL-1/CD162 Monoclonal antibody MAB996 Specifically detects PSGL-1/CD162 in Human samples. It is validated for Blocking (Adhesion), Flow Cytometry
Specifications
Specifications
PSGL-1/CD162
CHO131
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG
Mouse
IgM
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human PSGL-1/CD162. Recognizes sLe x -bearing core 2 O-glycan stuctures. It does not recognize sLe x on an extended core 1 O-glycan. The sLe x -bearing, core 2 O-glycan structure decorates the P-Selectin ligand PSGL-1, and the presence of this glycan structure is required for high affinity P-Selectin binding (1). This antibody stains human and canine leukocytes but does not recognize monkey, mouse, rabbit, porcine, feline or bovine leukocytes.
Human
Flow Cytometry, Adhesion Blockade, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade, CyTOF-ready
Affinity Purified
SELPLG
CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162
IgM-specific Affinity-purified from hybridoma culture supernatant
RUO
Primary
6404
Reconstitute at 0.5 mg/mL in sterile PBS.