missing translation for 'onlineSavingsMsg'
Learn More

PPP2R1A, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112384

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11525942

  • £257.00 / 100µg


Estimated Shipment
23-05-2018
Add to basket

Specifications

Specifications

Protein Phosphatase 2 A, A
Polyclonal
PBS with 0.08% sodium azide
Q32PI5, P30153, Q76MZ3
PP2Ac, PP2CA, PP2Calpha, RP-C
Rabbit
IgG
100μg
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
PPP2R1A
Purified
PPP2R1A
Synthetic peptide.
Ammonium sulfate precipitation
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
5518
Videos