missing translation for 'onlineSavingsMsg'
Learn More

Proliferin-related Protein/Plfr Rat anti-Mouse, Clone: 219310, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1527

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488705

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Proliferin-related Protein/Plfr Monoclonal antibody MAB1527 Specifically detects Proliferin-related Protein/Plfr in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Proliferin-related Protein/Plfr
219310
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Proliferin-related Protein/Plfr in direct ELISAs and Western blots.
Mouse
Western Blot
LYOPH
Western Blot 1 ug/mL
P04769
Prl7d1
Mouse myeloma cell line NS0-derived recombinant mouse Proliferin-related Protein/Plfr, Glu26-Phe244, Accession # P04769
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5617
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only