Learn More
Prolactin R Mouse anti-Rat, Clone: 218623, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB11121
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428355
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Prolactin R Monoclonal antibody MAB11121 Specifically detects Prolactin R in Rat samples. It is validated for Western Blotting
Specifications
Specifications
Prolactin R
218623
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PRLR
Mouse myeloma cell line NS0-derived recombinant rat Prolactin R Gln20-Asp229 Accession # P05710
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5618
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P05710
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRLR, PRL-R, prolactin receptor, prolactin receptor delta 7/11, ProlactinR, secreted prolactin binding protein
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat Prolactin R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Prolactin R or recombinant mouse Prolactin R is observed.
Rat