Learn More
Progranulin/PGRN Rat anti-Mouse, Clone: 333708, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2557
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439535
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
Progranulin/PGRN Monoclonal antibody MAB2557 Specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
Progranulin/PGRN
333708
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P28798
Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Progranulin/PGRN in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human Progranulin is observed.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Grn
Mouse myeloma cell line NS0-derived recombinant mouse Progranulin/PGRN Thr18-Leu589 Accession # P28798
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2896
Sterile PBS to a final concentration of 0.5 mg/mL.