missing translation for 'onlineSavingsMsg'
Learn More
Learn More
proBDNF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSB00016G
Code : LD
Additional Details : Weight : 0.99000kg
Description
Reconstitute with 500 μL of distilled water.
proBDNF plays a role in the development of hippocampal long term potentiation; involved in regulation of synaptic plasticity.Specifications
proBDNF | |
Polyclonal | |
Bdnf | |
Brain-derived neurotrophic factor precursor, proBDNF, pro BDNF | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
12064, 24225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P21237, P23363 | |
BDNF | |
A synthetic peptide from mouse pro BDNF conjugated to an immunogenic carrier protein was used as the antigen | |
500 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |