missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pro-Caspase 3 Monoclonal Antibody (31A893), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA141163
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: HL-60 or staurosporine treated apoptotic HeLa.
pro-Caspase 3 Monoclonal antibody specifically detects pro-Caspase 3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
pro-Caspase 3 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
P42574 | |
CASP3 | |
Recombinant full-length human Caspase-3. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
31A893 | |
Unconjugated | |
CASP3 | |
A830040C14Rik; AC-3; Apopain; Casp3; CASP-3; caspase 3; caspase 3, apoptosis related cysteine protease; caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CC3; Cpp32; CPP-32; CPP32B; cysteine protease CPP32; LICE; mLdy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama | |
Mouse | |
Protein G | |
RUO | |
836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |