Learn More
Presenilin-2 C-Terminal Fragment Mouse anti-Human, Clone: 149903, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1218
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488455
  • £358.00 / 500µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Presenilin-2 C-Terminal Fragment Monoclonal antibody MAB1218 Specifically detects Presenilin-2 C-Terminal Fragment in Human samples. It is validated for Western Blotting
Specifications
Specifications
Presenilin-2 C-Terminal Fragment
Unconjugated
Murine
IgG1
500μg
Primary
Human
149903
Purified
E.Coli-derived recombinant human Presenilin-2 C-Terminal Fragment. Glu322-Thr388
Protein A or G purified from hybridoma culture supernatant