Learn More
PRDC/GREM2 Rat anti-Mouse, Clone: 377407, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2069
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459145
  • £249.49 / 100µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
PRDC/GREM2 Monoclonal antibody MAB2069 Specifically detects PRDC/GREM2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
PRDC/GREM2
377407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GREM2
E. coli-derived recombinant mouse PRDC/GREM2 Arg22-Gln168 Accession # O88273
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
64388
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O88273
CKTSF1B2, CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3, DAND3Prdc, GREM2, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse PRDC/GREM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) DAN, rmCerberus, rmGremlin, or rmCoco is observed.