missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA515581
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
PPP3CB Polyclonal antibody specifically detects PPP3CB in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PPP3CB | |
Polyclonal | |
Unconjugated | |
PPP3CB | |
1110063J16Rik; calcineurin A; calcineurin A beta; Calcineurin A protein; calcineurin A2; Calcineurin subunit A beta; calmodulin-dependent calcineurin A subunit beta isoform; CALNA2; Calnb; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; CNA2; Cnab; CnAbeta; hypothetical protein LOC550554; PP2BA beta; PP2Bbeta; Ppp3cb; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3 catalytic subunit beta; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; RP11-345K20.1; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit alpha; wu:fq19c08; zgc:110141 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
19056, 5532 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P16298, P48453 | |
PPP3CB | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPP3CB | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |