Learn More
PP1 alpha Monoclonal Antibody (5E9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517155
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17155 targets PPP1CA in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17155 immunogen is purified recombinant fragment of human PPP1CA expressed in E. Coli. MA5-17155 detects PPP1CA which has a predicted molecular weight of approximately 38kDa.
PP1 alpha Monoclonal antibody specifically detects PP1 alpha in Human samples. It is validated for ELISA, Western Blot.Specifications
PP1 alpha | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P62136 | |
PPP1CA | |
Purified recombinant fragment of human PPP1CA expressed in E. Coli. | |
RUO | |
5499 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5E9 | |
Unconjugated | |
PPP1CA | |
96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |